siRNA Details
| Gene Symbol: Olfr504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011858.1 | NP_001011858.1 | 1 | 860 | ||
| GenBank mRNA | BC104310.1 | - | 1 | 860 | 40044370 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr504| Gene Symbol: Olfr504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011858.1 | NP_001011858.1 | 1 | 860 | ||
| GenBank mRNA | BC104310.1 | - | 1 | 860 | 40044370 | |