siRNA Details
| Gene Symbol: Olfr504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011858.1 | NP_001011858.1 | 1 | 694 | ||
| GenBank mRNA | BC104310.1 | - | 1 | 694 | 40044370 | |
| Gene Symbol: Olfr657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146312.2 | NP_666424.2 | 1 | 694 | ||
| GenBank mRNA | BC104311.1 | - | 1 | 689 | ||

Olfr504
Genomic Map