siRNA Details
| Gene Symbol: Olfr988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011534.1 | NP_001011534.1 | 1 | 133 | ||
| GenBank mRNA | BC150840.1 | - | 1 | 207 | 9087751 | |
| Gene Symbol: Olfr987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011785.1 | NP_001011785.1 | 1 | 133 | ||
| GenBank mRNA | BC147165.1 | - | 1 | 209 | 9056167 | |
| BC147166.1 | - | 1 | 209 | 9056185 | ||

Olfr988
Genomic Map