siRNA Details
| Gene Symbol: Olfr1182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011535.2 | NP_001011535.1 | 3 | 384 | ||
| GenBank mRNA | DR065631.1 | - | 3 | 384 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1182| Gene Symbol: Olfr1182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011535.2 | NP_001011535.1 | 3 | 384 | ||
| GenBank mRNA | DR065631.1 | - | 3 | 384 | ||