siRNA Details
| Gene Symbol: Olfr566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011536.1 | NP_001011536.1 | 1 | 664 | ||
| Gene Symbol: Olfr585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC145897.1 | - | 1 | 696 | 40131200 | |
More Information
Set Membership: |
|

Olfr566
Genomic Map