siRNA Details
| Gene Symbol: Olfr1532-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011542.1 | NP_001011542.1 | 1 | 221 | ||
| Gene Symbol: Olfr707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005570.2 | NP_001005570.2 | 1 | 221 | ||

Olfr1532-ps1
Genomic Map