siRNA Details
| Gene Symbol: Olfr406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011863.1 | NP_001011863.1 | 1 | 829 | ||
| GenBank mRNA | BC147149.1 | - | 1 | 876 | 9056162 | |
| BC147150.1 | - | 1 | 876 | 9056170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr406| Gene Symbol: Olfr406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011863.1 | NP_001011863.1 | 1 | 829 | ||
| GenBank mRNA | BC147149.1 | - | 1 | 876 | 9056162 | |
| BC147150.1 | - | 1 | 876 | 9056170 | ||