siRNA Details
| Gene Symbol: Olfr1328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146399.2 | NP_666511.2 | 1 | 830 | ||
| GenBank mRNA | BC106978.2 | - | 1 | 830 | 40042180 | |
| BC148290.2 | - | 1 | 830 | 40042169 | ||
| Gene Symbol: Olfr1329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011870.2 | NP_001011870.2 | 1 | 830 | ||

Olfr1328
Genomic Map