siRNA Details
| Gene Symbol: Olfr1034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011872.2 | NP_001011872.2 | 1 | 677 | ||
| GenBank mRNA | BC147677.1 | - | 1 | 677 | 9056650 | |
| Gene Symbol: Olfr1036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207142.2 | NP_997025.1 | 1 | 675 | ||
| GenBank mRNA | BC145856.1 | - | 1 | 675 | 40131159 | |

Olfr1034
Genomic Map