siRNA Details
| Gene Symbol: Olfr1034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011872.2 | NP_001011872.2 | 1 | 548 | ||
| GenBank mRNA | BC147677.1 | - | 1 | 548 | 9056650 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1034| Gene Symbol: Olfr1034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011872.2 | NP_001011872.2 | 1 | 548 | ||
| GenBank mRNA | BC147677.1 | - | 1 | 548 | 9056650 | |