siRNA Details
| Gene Symbol: Olfr102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011721.2 | NP_001011721.2 | 1 | 510 | ||
| GenBank mRNA | BC139114.1 | - | 1 | 510 | 8862136 | |
| BC139115.1 | - | 1 | 510 | 8862137 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr102| Gene Symbol: Olfr102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011721.2 | NP_001011721.2 | 1 | 510 | ||
| GenBank mRNA | BC139114.1 | - | 1 | 510 | 8862136 | |
| BC139115.1 | - | 1 | 510 | 8862137 | ||