siRNA Details
| Gene Symbol: Olfr1471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207132.2 | NP_997015.2 | 1 | 515 | ||
| Gene Symbol: Olfr1475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146301.1 | NP_666413.1 | 1 | 515 | ||

Olfr1471
Genomic Map