siRNA Details
| Gene Symbol: Olfr1061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207134.1 | NP_997017.1 | 1 | 658 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1061| Gene Symbol: Olfr1061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207134.1 | NP_997017.1 | 1 | 658 | ||