siRNA Details
| Gene Symbol: Olfr1084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207135.2 | NP_997018.2 | 1 | 600 | ||
| Gene Symbol: Olfr1080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146409.2 | NP_666521.2 | 2 | 625 | ||

Olfr1084
Genomic Map