siRNA Details
| Gene Symbol: Olfr417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207137.3 | NP_997020.3 | 1 | 477 | ||
| Gene Symbol: Olfr248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146714.2 | NP_666925.2 | 1 | 502 | ||
| GenBank mRNA | BC131960.1 | - | 1 | 502 | 40130237 | |
| BC131962.1 | - | 1 | 502 | 40130239 | ||
More Information
Set Membership: |
|

Olfr417
Genomic Map