siRNA Details
| Gene Symbol: Olfr576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001805.2 | NP_001001805.2 | 1 | 583 | ||
| GenBank mRNA | BC106977.2 | - | 1 | 583 | 40042102 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr576| Gene Symbol: Olfr576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001805.2 | NP_001001805.2 | 1 | 583 | ||
| GenBank mRNA | BC106977.2 | - | 1 | 583 | 40042102 | |