siRNA Details
| Gene Symbol: Olfr1308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207151.1 | NP_997034.1 | 1 | 905 | ||
| GenBank mRNA | BC108948.2 | - | 1 | 903 | ||
| BC108949.2 | - | 1 | 904 | |||
| BC115847.1 | - | 1 | 904 | |||
| BC115949.1 | - | 1 | 904 | 40043972 | ||

Olfr1308
Genomic Map