siRNA Details
| Gene Symbol: Olfr747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207156.2 | NP_997039.2 | 1 | 400 | ||
| Gene Symbol: Olfr749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020288.2 | NP_064684.2 | 2 | 466 | ||
More Information
Set Membership: |
|

Olfr747
Genomic Map