siRNA Details
| Gene Symbol: Olfr247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146269.2 | NP_666381.2 | 867 | |||
| Gene Symbol: Olfr819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165944.1 | NP_001159416.1 | 1 | 885 | ||

Olfr247
Genomic Map