siRNA Details
| Gene Symbol: Olfr448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146273.1 | NP_666385.1 | 1 | 210 | ||
| GenBank mRNA | BC107336.1 | - | 1 | 202 | ||
| BC107337.1 | - | 1 | 210 | 40055003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr448| Gene Symbol: Olfr448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146273.1 | NP_666385.1 | 1 | 210 | ||
| GenBank mRNA | BC107336.1 | - | 1 | 202 | ||
| BC107337.1 | - | 1 | 210 | 40055003 | ||