siRNA Details
| Gene Symbol: Olfr284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146281.1 | NP_666393.1 | 1 | 637 | ||
| GenBank mRNA | BC111907.1 | - | 1 | 626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr284| Gene Symbol: Olfr284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146281.1 | NP_666393.1 | 1 | 637 | ||
| GenBank mRNA | BC111907.1 | - | 1 | 626 | ||