siRNA Details
| Gene Symbol: Olfr846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146282.2 | NP_666394.1 | 1 | 855 | ||
| GenBank mRNA | BC134375.1 | - | 1 | 856 | 40135046 | |
| Gene Symbol: Olfr847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146525.1 | NP_666736.1 | 1 | 852 | ||
| GenBank mRNA | BC111580.1 | - | 1 | 851 | ||

Olfr846
Genomic Map