siRNA Details
| Gene Symbol: Olfr846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146282.2 | NP_666394.1 | 1 | 235 | ||
| GenBank mRNA | BC134375.1 | - | 1 | 236 | 40135046 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr846| Gene Symbol: Olfr846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146282.2 | NP_666394.1 | 1 | 235 | ||
| GenBank mRNA | BC134375.1 | - | 1 | 236 | 40135046 | |