siRNA Details
| Gene Symbol: Olfr114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146287.1 | NP_666399.1 | 1 | 478 | ||
| GenBank mRNA | BC132209.1 | - | 1 | 508 | 40130486 | |
| BC132573.1 | - | 1 | 508 | 40130850 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr114| Gene Symbol: Olfr114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146287.1 | NP_666399.1 | 1 | 478 | ||
| GenBank mRNA | BC132209.1 | - | 1 | 508 | 40130486 | |
| BC132573.1 | - | 1 | 508 | 40130850 | ||