siRNA Details
| Gene Symbol: Olfr122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146288.3 | NP_666400.3 | 2 | 207 | ||
| GenBank mRNA | CB173933.1 | - | 2 | 207 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr122| Gene Symbol: Olfr122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146288.3 | NP_666400.3 | 2 | 207 | ||
| GenBank mRNA | CB173933.1 | - | 2 | 207 | ||