siRNA Details
| Gene Symbol: Olfr121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146629.2 | NP_666840.2 | 2 | 612 | ||
| GenBank mRNA | BC090631.1 | - | 2 | 612 | 30358555 | |
| Gene Symbol: Olfr122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146288.3 | NP_666400.3 | 2 | 588 | ||

Olfr121
Genomic Map