siRNA Details
| Gene Symbol: Olfr113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146289.1 | NP_666401.1 | 1 | 770 | ||
| GenBank mRNA | BC132178.1 | - | 1 | 797 | 40130455 | |
| BC132180.1 | - | 1 | 797 | 40130457 | ||
| Gene Symbol: Olfr114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146287.1 | NP_666399.1 | 1 | 770 | ||
| GenBank mRNA | BC132209.1 | - | 1 | 800 | 40130486 | |
| BC132573.1 | - | 1 | 800 | 40130850 | ||

Olfr113
Genomic Map