siRNA Details
| Gene Symbol: Olfr1143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146293.2 | NP_666405.1 | 1 | 334 | ||
| GenBank mRNA | CB173239.1 | - | 1 | 420 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1143| Gene Symbol: Olfr1143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146293.2 | NP_666405.1 | 1 | 334 | ||
| GenBank mRNA | CB173239.1 | - | 1 | 420 | ||