siRNA Details
| Gene Symbol: Olfr1167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146294.2 | NP_666406.2 | 1 | 534 | ||
| GenBank mRNA | BC119292.1 | - | 1 | 534 | 8734041 | |
| BC137817.1 | - | 1 | 534 | 8860836 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1167| Gene Symbol: Olfr1167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146294.2 | NP_666406.2 | 1 | 534 | ||
| GenBank mRNA | BC119292.1 | - | 1 | 534 | 8734041 | |
| BC137817.1 | - | 1 | 534 | 8860836 | ||