siRNA Details
| Gene Symbol: Olfr1115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146297.2 | NP_666409.2 | 1 | 550 | ||
| GenBank mRNA | BC141024.1 | - | 1 | 550 | 9055636 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1115| Gene Symbol: Olfr1115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146297.2 | NP_666409.2 | 1 | 550 | ||
| GenBank mRNA | BC141024.1 | - | 1 | 550 | 9055636 | |