siRNA Details
| Gene Symbol: Olfr1475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146301.1 | NP_666413.1 | 1 | 631 | ||
| Gene Symbol: Olfr1472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146690.2 | NP_666901.2 | 1 | 631 | ||

Olfr1475
Genomic Map