siRNA Details
| Gene Symbol: Olfr1340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146304.2 | NP_666416.2 | 1 | 351 | ||
| GenBank mRNA | BC106795.1 | - | 1 | 351 | 40042190 | |
| BC126866.1 | - | 1 | 345 | |||
| BC126867.1 | - | 1 | 346 | 40042921 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1340| Gene Symbol: Olfr1340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146304.2 | NP_666416.2 | 1 | 351 | ||
| GenBank mRNA | BC106795.1 | - | 1 | 351 | 40042190 | |
| BC126866.1 | - | 1 | 345 | |||
| BC126867.1 | - | 1 | 346 | 40042921 | ||