siRNA Details
Gene Symbol: Olfr420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146305.2 | NP_666417.2 | 1 | 266 | ||
GenBank mRNA | BC145885.1 | - | 1 | 266 | 40131188 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146305.2 | NP_666417.2 | 1 | 266 | ||
GenBank mRNA | BC145885.1 | - | 1 | 266 | 40131188 |