siRNA Details
| Gene Symbol: Olfr498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146307.2 | NP_666419.2 | 1 | 253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr498| Gene Symbol: Olfr498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146307.2 | NP_666419.2 | 1 | 253 | ||