siRNA Details
| Gene Symbol: Olfr601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146314.2 | NP_666426.2 | 1 | 880 | ||
| GenBank mRNA | BC131956.1 | - | 1 | 880 | 40130233 | |
| BC131958.1 | - | 1 | 880 | 40130235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr601| Gene Symbol: Olfr601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146314.2 | NP_666426.2 | 1 | 880 | ||
| GenBank mRNA | BC131956.1 | - | 1 | 880 | 40130233 | |
| BC131958.1 | - | 1 | 880 | 40130235 | ||