siRNA Details
| Gene Symbol: Olfr601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146314.2 | NP_666426.2 | 1 | 544 | ||
| GenBank mRNA | BC131956.1 | - | 1 | 544 | 40130233 | |
| BC131958.1 | - | 1 | 544 | 40130235 | ||
| Gene Symbol: Olfr605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011854.2 | NP_001011854.2 | 1 | 552 | ||
| GenBank mRNA | BC147216.1 | - | 1 | 552 | 9056257 | |
| BC147217.1 | - | 1 | 552 | 9056245 | ||

Olfr601
Genomic Map