siRNA Details
Gene Symbol: Olfr186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146321.1 | NP_666433.1 | 1 | 229 | ||
GenBank mRNA | BC150713.1 | - | 1 | 251 | 9087624 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146321.1 | NP_666433.1 | 1 | 229 | ||
GenBank mRNA | BC150713.1 | - | 1 | 251 | 9087624 |