siRNA Details
| Gene Symbol: Olfr809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146324.1 | NP_666436.1 | 1 | 311 | ||
| GenBank mRNA | BC120625.1 | - | 1 | 355 | 40129548 | |
| BC137816.1 | - | 1 | 355 | 8860835 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr809| Gene Symbol: Olfr809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146324.1 | NP_666436.1 | 1 | 311 | ||
| GenBank mRNA | BC120625.1 | - | 1 | 355 | 40129548 | |
| BC137816.1 | - | 1 | 355 | 8860835 | ||