siRNA Details
Gene Symbol: Olfr111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005485.2 | NP_001005485.1 | 1 | 788 | ||
GenBank mRNA | BC139116.1 | - | 1 | 788 | 8862138 | |
BC139117.1 | - | 1 | 788 | 8862139 |
Gene Symbol: Olfr110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146328.2 | NP_666440.2 | 2 | 850 | ||
GenBank mRNA | BC132347.1 | - | 1 | 794 | 40130624 |
