siRNA Details
| Gene Symbol: Olfr954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146331.1 | NP_666443.1 | 1 | 587 | ||
| Gene Symbol: Olfr951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011812.1 | NP_001011812.1 | 1 | 587 | ||

Olfr954
Genomic Map