siRNA Details
| Gene Symbol: Olfr135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146332.1 | NP_666444.1 | 1 | 766 | ||
| GenBank mRNA | BC116774.1 | - | 1 | 844 | 40126093 | |
| BC116776.1 | - | 1 | 844 | 40126095 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr135| Gene Symbol: Olfr135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146332.1 | NP_666444.1 | 1 | 766 | ||
| GenBank mRNA | BC116774.1 | - | 1 | 844 | 40126093 | |
| BC116776.1 | - | 1 | 844 | 40126095 | ||