siRNA Details
| Gene Symbol: Olfr135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146332.1 | NP_666444.1 | 1 | 240 | ||
| GenBank mRNA | BC116774.1 | - | 1 | 318 | 40126093 | |
| BC116776.1 | - | 1 | 318 | 40126095 | ||
| Gene Symbol: Olfr133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146831.1 | NP_667042.1 | 1 | 240 | ||
| GenBank mRNA | BC132057.1 | - | 1 | 329 | 40130334 | |
| BC132059.1 | - | 1 | 329 | 40130336 | ||

Olfr135
Genomic Map