siRNA Details
Gene Symbol: Olfr1330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146334.2 | NP_666446.1 | 1 | 841 | ||
GenBank mRNA | BC108943.1 | - | 1 | 841 | 40042936 | |
BC108944.1 | - | 1 | 836 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146334.2 | NP_666446.1 | 1 | 841 | ||
GenBank mRNA | BC108943.1 | - | 1 | 841 | 40042936 | |
BC108944.1 | - | 1 | 836 |