siRNA Details
Gene Symbol: Olfr893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146336.2 | NP_666448.2 | 1 | 287 | ||
GenBank mRNA | BC130242.1 | - | 1 | 288 | 40142233 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146336.2 | NP_666448.2 | 1 | 287 | ||
GenBank mRNA | BC130242.1 | - | 1 | 288 | 40142233 |