siRNA Details
| Gene Symbol: Olfr1396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146337.1 | NP_666449.1 | 1 | 70 | ||
| GenBank mRNA | BC110470.1 | - | 1 | 67 | 40044195 | |
| BC110471.1 | - | 1 | 70 | 40044201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1396| Gene Symbol: Olfr1396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146337.1 | NP_666449.1 | 1 | 70 | ||
| GenBank mRNA | BC110470.1 | - | 1 | 67 | 40044195 | |
| BC110471.1 | - | 1 | 70 | 40044201 | ||