siRNA Details
| Gene Symbol: Olfr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146339.3 | NP_666451.1 | 3 | 1194 | ||
| GenBank mRNA | BC046590.1 | - | 3 | 1194 | 6309128 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr77| Gene Symbol: Olfr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146339.3 | NP_666451.1 | 3 | 1194 | ||
| GenBank mRNA | BC046590.1 | - | 3 | 1194 | 6309128 | |