siRNA Details
| Gene Symbol: Olfr1259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146341.1 | NP_666453.1 | 1 | 669 | ||
| Gene Symbol: Olfr1255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146977.2 | NP_667188.2 | 1 | 687 | ||

Olfr1259
Genomic Map