siRNA Details
Gene Symbol: Olfr1269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146342.1 | NP_666454.1 | 1 | 74 | ||
GenBank mRNA | BC116956.1 | - | 1 | 95 | 40126275 | |
BC116958.1 | - | 1 | 95 | 40126277 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146342.1 | NP_666454.1 | 1 | 74 | ||
GenBank mRNA | BC116956.1 | - | 1 | 95 | 40126275 | |
BC116958.1 | - | 1 | 95 | 40126277 |