siRNA Details
| Gene Symbol: Olfr1491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146345.1 | NP_666457.1 | 1 | 227 | ||
| GenBank mRNA | BC128268.1 | - | 1 | 227 | 40043876 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1491| Gene Symbol: Olfr1491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146345.1 | NP_666457.1 | 1 | 227 | ||
| GenBank mRNA | BC128268.1 | - | 1 | 227 | 40043876 | |