siRNA Details
| Gene Symbol: Olfr397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146346.1 | NP_666458.1 | 1 | 587 | ||
| GenBank mRNA | BC120574.1 | - | 1 | 667 | 40129497 | |
| BC125526.1 | - | 1 | 667 | 40130041 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr397| Gene Symbol: Olfr397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146346.1 | NP_666458.1 | 1 | 587 | ||
| GenBank mRNA | BC120574.1 | - | 1 | 667 | 40129497 | |
| BC125526.1 | - | 1 | 667 | 40130041 | ||